Anti-UBQLN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
23372-1-AP-150UL
10081-320EA 462.47 USD
10081-320
Anti-UBQLN3 Rabbit Polyclonal Antibody
Antibodies
UBQLN3(Ubiquilin-3) may interact with the testis-specific cyclin A1 and regulate cell cycle progression during spermatogenesis, as does frog Xdrp1, with which UBQLN3 shares sequence similarity.UBQLN3 expression is not observed in mouse eye, thyroid, pancreas,smooth muscle, ovary, submaxillary gland, uterus, and epididymis by northern blot.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: UBQLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR