Anti-RBM17 Rabbit Polyclonal Antibody

Supplier: Proteintech
13918-1-AP-150UL
10081-266EA 462.47 USD
10081-266
Anti-RBM17 Rabbit Polyclonal Antibody
Antibodies
Western Blot:BxPC-3 Cells, 1:200-1:2000; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: RBM17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR