Anti-TTF1 Mouse Monoclonal Antibody [clone: 2C8F3]

Supplier: Proteintech
66034-1-IG-150UL
10081-256EA 462.47 USD
10081-256
Anti-TTF1 Mouse Monoclonal Antibody [clone: 2C8F3]
Antibodies
Homeobox protein Nkx-2.1 (NKX2-1), also named Thyroid nuclear factor 1,Thyroid transcription factor 1(TTF1). It is a transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. Crucial in the maintenance of the thyroid differentiation phenotype. May play a role in lung development and surfactant homeostasis. Defects in NKX2-1 are the cause of benign hereditary chorea (BHC), choreoathetosis, hypothyroidism, and neonatal respiratory distress (CHNRD).

Western Blot: Hela Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TTF1
Clonality: Monoclonal
Clone: 2C8F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR