Anti-transcription termination factor-Specific Mouse Monoclonal Antibody [clone: 7B3H2]

Supplier: Proteintech
60235-1-IG-150UL
10081-254EA 462.47 USD
10081-254
Anti-transcription termination factor-Specific Mouse Monoclonal Antibody [clone: 7B3H2]
Antibodies
TTF1 is a multifunctional nucleolar protein that terminates ribosomal gene transcription, mediates replication fork arrest and regulates RNA polymerase I transcription on chromatin. It plays a dual role in rDNA regulation, being involved in both activation and silencing of rDNA transcription. Interaction with BAZ2A/TIP5, TTF1 recovers DNA-binding activity. The antibody is specific to TTF1.

Western Blot: Hela cell, 1:1000-1:10000

Type: Primary
Antigen: transcription termination factor-Specific
Clonality: Monoclonal
Clone: 7B3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR