Anti-ACVRL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14745-1-AP-150UL
10081-250EA 462.47 USD
10081-250
Anti-ACVRL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000; IHC: Human Lymphoma Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: ACVRL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR