Anti-PSAP Rabbit Polyclonal Antibody

Supplier: Proteintech
18396-1-AP-150UL
10081-200EA 462.47 USD
10081-200
Anti-PSAP Rabbit Polyclonal Antibody
Antibodies
The PSAP gene encodes prosaposin, a precursor of fourl small nonenzymatic glycoproteins termed 'sphingolipid activator proteins' (SAPs) that assist in the lysosomal hydrolysis of sphingolipids. After proteolytic processing of the presaposin protein, these 4 released polypeptides are functional activators. Saposin A was encoded by residues 60 to 143 of PSAP, saposin B by 195 to 275, saposin C by 311 to 390, and saposin D by 405 to 487. They are four 12-14 kDa heatstable glycoproteins. This antibody should recognize saposin A.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR