Anti-TRPS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21938-1-AP-150UL
10081-192EA 462.47 USD
10081-192
Anti-TRPS1 Rabbit Polyclonal Antibody
Antibodies
TRPS1 is a zinc finger transcriptional repressor involved in the regulation of chondrocyte and perichondrium development, containing a GATA-type zinc finger through which it binds to DNA. with nine zinc-finger domains and two C-terminal Ikaros-like zinc fingers. Its reperssible function was dependent on the integrity of the Trps1 GATA-type zinc-finger domain and also required the C-terminal 119 amino acids of the protein, which harbor the two Ikaros-like zinc-finger domains

Western Blot: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: TRPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR