Anti-Troponin I Type 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22130-1-AP-150UL
10081-180EA 462.47 USD
10081-180
Anti-Troponin I Type 2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: Troponin I Type 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR