Anti-TLE3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22094-1-AP-150UL
10081-168EA 462.47 USD
10081-168
Anti-TLE3 Rabbit Polyclonal Antibody
Antibodies
Transducin-like enhancer of split 3(TLE3) is a corepressor that binds to a variant of transciption factor, which mediated by CTNNB1 and TCF family members in Wnt signaling. It's suggested that effect of full-length TLE family members be regulated by association with dominant-negative AES.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TLE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR