Anti-TLE3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11372-1-AP-150UL
10081-166EA 462.47 USD
10081-166
Anti-TLE3 Rabbit Polyclonal Antibody
Antibodies
TLE3, also named as KIAA1547 and ESG3, belongs to the WD repeat Groucho/TLE family. It is a transcriptional corepressor that binds to a number of transcription factors. It inhibits the transcriptional activation mediated by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES. TLE3 is a member of the transducin-like enhancer of split (TLE) family of proteins that have been implicated in the tumorgenesis and classification of sarcomas. It is a candidate biomarker of response to taxane

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: TLE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR