Anti-SYK Rabbit Polyclonal Antibody

Supplier: Proteintech
14858-1-AP-150UL
10081-136EA 462.47 USD
10081-136
Anti-SYK Rabbit Polyclonal Antibody
Antibodies
SYK belongs to the protein kinase superfamily, Tyr protein kinase family and SYK/ZAP-70 subfamily. It is positive effector of BCR-stimulated responses. SYK couples the B-cell antigen receptor (BCR) to the mobilization of calcium ion either through a phosphoinositide 3-kinase-dependent pathway, when not phosphorylated on tyrosines of the linker region, or through a phospholipase C-gamma-dependent pathway It phosphorylates USP25 and regulates its intracellular levels.

IHC: Human spleen, 1:20-1:200

Type: Primary
Antigen: SYK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR