Battery Backpack Vac, Atrix
Supplier: Atrix International
VACBP36V
100720-986EA 1285 USD
100720-986
Battery Backpack Vac, Atrix
Vacuum Cleaners
The Atrix Battery Backpack is perfect for cleaning confined areas and where corded electric vacuums are not a viable option
The Atrix International VACBP6-5P HEPA Replacement Filter Bags are designed to capture particulates as small as 0.3 microns when using the Backpack Series Vacuum Systems.
The battery feature eliminates all safety hazards that corded equipment may cause. Users are granted the freedom to clean anywhere such as on scaffolding, entry ways, attics, or in stairwells. Ergonomic and comfortable fit for all sizes. The VACBP36V has a continuous run time of 50-55 minutes andMore Product Information