Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Prosci
35-318
10071-992EA 633.48 USD
10071-992
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.

Mycoplasma pneumoniae antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Mycoplasma pneumoniae Antibody (FITC)
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR