HyperREZ™ XP Guard Cartridge Holder, Thermo Scientific
Supplier: Thermo Scientific
60002-354
10067-730EA 550.65 USD
10067-730
HyperREZ™ XP Guard Cartridge Holder, Thermo Scientific
Guard Column Holders
Choose the Thermo Scientific HyperREZ XP Guard Cartridge Holder for use with HyperREZ XP 3.0 x 5.0 mm Guard Cartridges.