Human Fibroblast Growth Factor 7
Supplier: Thermo Scientific Chemicals
J67052-EXE
10067-302EA
120.84
USD
10067-302
10067-304
Human Fibroblast Growth Factor 7
Proteins and Peptides
Fibroblast Growth Factor 7, human
Formula Weight: 18.9 kDa
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00212245
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...