Cefoxitin sodium 92.7-97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
A broad spectrum antibiotic
Purity: 92.7-97%
CAS No.: 33564-30-6
Molecular Formula: C16H16N3NaO7S2
Formula Weight: 449.43
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: A broad spectrum antibiotic
Formula:
C₁₆H₁₆N₃NaO₇S₂ MW: 449.428 g/mol Boiling Pt: 843 °C Melting Pt: >160 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00079042 CAS Number: 33564-30-6 EINECS: 251-574-6 Merck Index: 14,1936 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...