oxolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Targets Gram-negative bacteria
Purity: 98%
CAS No.: 14698-29-4
Molecular Formula: C13H11NO5
Formula Weight: 261.23
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: Targets gram negative bacteria
Formula:
C₁₃H₁₁NO₅ Boiling Pt: 473 °C Melting Pt: 314…316 °C (dec.) Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00056775 CAS Number: 14698-29-4 EINECS: 238-750-8 Merck Index: 14,6965 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...