Chlorimuron ethyl 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J66605-03
J66605-06
10067-216EA
34.2
USD
10067-216
10067-218
10067-220
Chlorimuron ethyl 95%
Chlorimuron ethyl
Chlorimuron ethyl, 95%
Purity: 95%
CAS No.: 90982-32-4
Molecular Formula: C15H15ClN4O6S
Formula Weight: 414.82
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Colorless
Melting Point: 180-182°
Solubility: Freely soluble in aqueous solution
MDL No.: MFCD00128063
Formula:
C₁₅H₁₅ClN₄O₆S MW: 414.817 g/mol Melting Pt: 180…182 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00128063 CAS Number: 90982-32-4 Merck Index: 14,2093 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...