Test Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
103101 112201 115261 120161 120180 136101 151101 160101 160201 163161 163177 163261 164161 169101 187261 187262 191161 191180 186171 188161 188171 115101 115262 187101 187201 163201
82050-104CS 144.03 USD
82050-104 82050-116 82050-138 82050-148 82050-150 82050-228 82050-232 82050-234 82050-238 82050-244 82050-246 82050-248 82050-252 82050-256 82050-266 82050-272 82050-274 82050-292 82050-294 89049-098 89049-100 89049-102 82050-128 82050-140 82050-268 82050-270 10066-770
Test Tubes, Greiner Bio-One
Tubes Culture Tubes
These high-quality test tubes are available in a wide variety of materials, sizes, and volume ranges.
  • Excellent optical clarity
  • Round, flat, or conical bottom designs available
  • Polycarbonate, polypropylene, or polystyrene construction available
  • Exceptional thermal and chemical stability
Test tubes provide excellent optical clarity, and are available in round or conical bottom designs. Unless otherwise noted, tube caps are not included.
Ordering information: Test tube 82050-248 is supplied sterile.