YSI MultiLab BOD Lab Kits, YSI
Supplier: YSI
626570
10066-708EA 3568.02 USD
10066-708 10066-710
YSI MultiLab BOD Lab Kits, YSI
Electrochemistry Meters pH and Multi-Parameter Meters
The YSI MultiLab line includes the 4010-1 (single channel), 4010-2 (dual channel) and 4010-3 (three channel) instruments providing easy to use and calibrate menu-driven operation ideal for the laboratory
  • One channel input for pH, ORP, DO/BOD, or conductivity
  • 500 data sets in manual mode and 5,000 data sets in automatic logging mode
  • Large, easy to read graphic display
  • Intelligent, digital sensors (plug and play sensors)
  • GLP traceability (sensors store serial # and calibration data)
  • USB connectivity to manage data
  • Includes self-stirring BOD, pH, ORP and conductivity sensors
Measure pH, ORP, conductivity or BOD accurately in the laboratory with the YSI 4010-1. This instrument has one channel to accommodate and of the probe choices. The IDS sensors automatically store their unique serial number and calibration data. In addition, they also digitally process the measurement signal. The sensors can be moved from instrument to instrument and maintain their calibration data and transmit this information to the new instrument. Plug and play connectivity with each instrument
Certifications: GLP compliant.
Ordering information: Sensors come with a three year warranty.