Tweezers, Straight Delrin Soft Replaceable Tips, Excelta Corp®
Supplier: Excelta
159B-RTWX 159D-RTWX 159C-RTWX
94024-106EA 11.63 USD
94024-106 94024-116 10066-508 76243-388
Tweezers, Straight Delrin Soft Replaceable Tips, Excelta Corp®
Forceps
These soft replaceable tip tweezers feature Acetal tips which are electrically insulative and resistant to temperatures up to 185 °F.
The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). Excellent for cleanroom applications and for avoiding the marring or scratching of items being handled.
All replacement tips come complete with 2 tips and 4 screws.