Tri-Sil BP (BSA:Pyridine) Reagent
Supplier: Thermo Scientific
TS-49012
10066-358EA 1041.91 USD
10066-358
Tri-Sil BP (BSA:Pyridine) Reagent
Chromatography Sorbents GC Sorbents
Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent, Composed of BSA and high-purity pyridine, this silylation reagent formulation derivatizes alcohols, phenols, organic acids, aromatic amides, and amines for GC analysis. It is excellent for use with unhindered steriods.
  • Standard components are Pyridine, Trimethylsilyl N-trimethylsilylacetamidate
  • Standard Concentration:65 - 80, 20 - 25%
  • For use with GC
  • Organic acids to form trimethylsilyl esters
  • Not recommended for use with carbohydrates