Methoxamine (MOX) Reagent, Thermo Scientific
Supplier: Thermo Scientific
TS-45950
10066-332EA 137.49 USD
10066-332
Methoxamine (MOX) Reagent, Thermo Scientific
Chromatography Sorbents GC Sorbents
MOX Reagent (M.W
  • Prevent multiple derivative formation
  • Used primarily with steroids in chromatography
It prevents the formation of multiple derivatives when enols are present during silylation. It has 2% solution of methoxyamine.HCI in pyridine, and it is used primarily with steroids in chromatography.
83.51) converts keto groups to methoxime derivatives.