Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
D3-113-FC
10065-878EA 166.07 USD
10065-878
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR