Anti-IgG2a Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-112-2A
10065-846EA 340.48 USD
10065-846
Anti-IgG2a Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR