Anti-IgM Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-110-M-100
10065-834EA 171.06 USD
10065-834 10065-836
Anti-IgM Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR