Anti-6xHis Mouse Monoclonal Antibody [clone: AD1.1.10] (HRP (Horseradish Peroxidase))

Supplier: Columbia Biosciences
HRP-1711
10065-762EA 570.46 USD
10065-762
Anti-6xHis Mouse Monoclonal Antibody [clone: AD1.1.10] (HRP (Horseradish Peroxidase))
Antibodies
Mouse Monocolonal [AD1.1.10] to 6X His tag (HRP)

Type:
Antigen: 6xHis
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR