Anti-YPYDVPDYA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 16B12]

Supplier: Columbia Biosciences
HRP-1722
10065-758EA 563.61 USD
10065-758
Anti-YPYDVPDYA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 16B12]
Antibodies
Type:
Antigen: YPYDVPDYA
Clonality: Monoclonal
Clone: 16B12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR