Anti-GKPIPNPLLGLDST Rabbit Polyclonal Antibody (DyLight® 550)

Supplier: Columbia Biosciences
D9-1834
10065-742EA 547.62 USD
10065-742
Anti-GKPIPNPLLGLDST Rabbit Polyclonal Antibody (DyLight® 550)
Antibodies
Type:
Antigen: GKPIPNPLLGLDST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR