Anti-EQKLISEEDL Mouse Monoclonal Antibody (SureLight® P3) [clone: 9E10]

Supplier: Columbia Biosciences
D7-1714
10065-698EA 547.62 USD
10065-698
Anti-EQKLISEEDL Mouse Monoclonal Antibody (SureLight® P3) [clone: 9E10]
Antibodies
Type:
Antigen: EQKLISEEDL
Clonality: Monoclonal
Clone: 9E10
Conjugation: SureLight® P3
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: c-myc tagged proteins
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR