Anti-YPYDVPDYA Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 16B12]

Supplier: Columbia Biosciences
D5-1722
10065-658EA 701.23 USD
10065-658
Anti-YPYDVPDYA Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 16B12]
Antibodies
Mouse monoclonal [16B12} to HA tag conjugated to SureLight R-PE

Type:
Antigen: YPYDVPDYA
Clonality: Monoclonal
Clone: 16B12
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR