Anti-EQKLISEEDL Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 9E10)]

Supplier: Columbia Biosciences
D5-1714
10065-654EA 563.61 USD
10065-654
Anti-EQKLISEEDL Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 9E10)]
Antibodies
Type:
Antigen: EQKLISEEDL
Clonality: Monoclonal
Clone: 9E10
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: c-myc tagged proteins
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR