Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
D3-110-M
10065-582EA 340.48 USD
10065-582
Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR