Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (Biotin)

Supplier: Columbia Biosciences
B1-1832
10065-470EA 563.61 USD
10065-470
Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MASMTGGQQMG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR