Anti-KETAAAKFERQHMDS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Columbia Biosciences
AP-1831
10065-454EA 563.61 USD
10065-454
Anti-KETAAAKFERQHMDS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type:
Antigen: KETAAAKFERQHMDS
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR