Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Columbia Biosciences
AP-1310
10065-444EA 563.61 USD
10065-444
Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR