Anti-CD8 T Cell Mouse Monoclonal Antibody [clone: 144B]
Supplier: Diagnostic Biosystems
MOB117 MOB117-05
99990-812EA 1173.52 USD
99990-812 10064-920
Anti-CD8 T Cell Mouse Monoclonal Antibody [clone: 144B]
Antibodies
This antibody reacts specifically with a 32 kDa protein similar to the CD8 molecule
  • Concentrated
This antibody stains cytotoxic/suppressor T cells and shows staining patterns similar to clustered CD8 monoclonal antibodies