Potassium Hydroxide, 10% KOH, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z77
10063-758EA 29.17 USD
10063-758
Potassium Hydroxide, 10% KOH, Hardy Diagnostics
Microbiology Test Systems Enzymatic Tests
For use in slide preparations of specimens for the purpose of identifying fungal elements.
  • Dissolves epithelial cells
For use in slide preparations of specimens for the purpose of identifying fungal elements
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Dissolves epithelial cells that may mask fungal elements.