AnaeroGen™ Compact System, Hardy Diagnostics
Supplier: Hardy Diagnostics
AN010C
10063-746EA 190.76 USD
10063-746
AnaeroGen™ Compact System, Hardy Diagnostics
Anaerobic Systems
AnaeroGen™ Compact is a system for generating an anaerobic environment rapidly to cultivate anaerobic microorganisms.
  • No catalyst needed and no water required for activation
  • No potentially explosive hydrogen gas produced
  • No dangerous build-up of pressure
  • Sachet is activated upon opening
  • Final atmosphere generated contains less than 1% oxygen, which is ideal for the growth of anaerobes
It consists of an AnaeroGen™ sachet, and oxygen impermeable pouch. Each pouch is large enough to hold four 15 × 100 mm petri plates.