SmartWrite Slide Printer, Sakura Finetek
Supplier: Sakura Finetek
9021
10062-828EA 21557.98 USD
10062-828
SmartWrite Slide Printer, Sakura Finetek
Printers Cassette and Slide Printers
Microscope slide printer with thermal transfer print technology, with multiple color or black ribbons
Prints directly on white 3 x 1" standard or positive charged slides with smooth surfaced white frosted end and clipped corners.
SwartWrite Software (9025), PC with touch screen (9026) and 2-D bar code scanner (9024) are all sold separately.
Data interface USB 2.0. Slide cartridge capacity 100 slides with slide output tray capacity 15 slides. Print resolution 300 dpi. Printer drivers are Windows XP & Windows 7 compatible. Print speed up to 9 slides/minute using black; up to 5 slides/minute using solid colors; up to 4 slides/minute using blended colors.
Scannable 2D and 1D bar Code, alphanumeric and graphic printing capabilities.
Ordering information: Warranty is one year, parts and labor.