Anti-NCBP1 Rabbit Polyclonal Antibody

Supplier: Prosci
7395
10061-918EA 580.46 USD
10061-918
Anti-NCBP1 Rabbit Polyclonal Antibody
Antibodies
NCBP1 Antibody: NCBP1, also known as CBP80, is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. NCBP1 promotes high-affinity mRNA-cap binding and associates with the CTD of RNA polymerase II. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay (1,2). Recent evidence has shown that cellular-cap-binding proteins such as NCBP1 associate with influenza virus mRNAs, suggesting that these viral mRNAs may follow the normal cellular pathways for splicing, nuclear export, and translation.

NCBP1 antibody can be used for detection of NCBP1 by Western blot at 1 - 2 ?g/mL.

NCBP1 Antibody is affinity chromatography purified via peptide column.

NCBP1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NCBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR