Level Sensors, S.C.A.T.
Supplier: S.C.A.T.
107881
10060-770EA 567.43 USD
10060-770 10060-772 10060-774 10060-964 10060-966 10060-968
Level Sensors, S.C.A.T.
Level Sensors
These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own weight while the barrel fills up.
  • For barrel opening with Ø 50 to 90 mm
  • Practical handhold
  • Easy to use with gloves
PFA float, PTFE/PE indicator
These level sensors are a reliable protection against overflow during the filling.