Safety Funnel Disposal Units, S.C.A.T.
Supplier: S.C.A.T.
107416
10060-944EA 766.46 USD
10060-944 10060-946 10060-948 10060-950
Safety Funnel Disposal Units, S.C.A.T.
Funnels Safety Funnels
Sets offer a way to get your waste fluid collection site ready for operation quickly. Kits come with grounding cable, sieve, and are PE-HD Electrically Conductive. A 10L container that is also PE-HD Electricallly Conductive is included in each kit. Ball Valve Funnel Kits have spalsh protection. Floats are for level control.
  • Only one part number for the comprehensive safety system
  • No time consuming compilation of a configuration
  • Price advantage compared to ordering piece by piece
  • The delivery is made complete – the system is immediately ready for use