Canisters with Integrated Floater, S.C.A.T.
Supplier: S.C.A.T.
108320
10060-768EA 352.52 USD
10060-768 10060-818 10060-820 10060-842 10061-002 10061-012 10002-746
Canisters with Integrated Floater, S.C.A.T.
Bottles Jerricans
Safely collect fluid laboratory waste. Ideal for use with S.C.A.T safety funnels.
The integrated floater gives a warning in time to prevent overfilling. Container from electrically conductive plastic counteracts electrostatic discharge and can be grounded. Thus they are especially well-suited for use in protected areas. Viewing Strip option has UN-Y approval.