Dissection Tray, AIMS™
Supplier: Aims
DTW1
10060-188EA 54.09 USD
10060-188
Dissection Tray, AIMS™
Trays/Dishes/Pots Dissecting Trays
Blue wax dissection tray is formulated to provide proper consistency to allow pins to stick firmly and be easily removed.
  • Uses a completely environmentally safe wax product
  • Surface can be renewed by heating and the dissection tray can be reused several times
The MSDS sheet for this product lists no toxic chemicals and there are no fire, health or reactivity hazards with this product.