AIMS™ Stereotaxic Magnifier
Supplier: Aims
SMAG
10060-178EA 299.8 USD
10060-178
AIMS™ Stereotaxic Magnifier
Magnifiers
Unique magnetic base/puck system allows the magnifier to attach to non-metallic surfaces.
  • Flex arm, non-obstructive rimless magnifying glass
  • Main lens: 2.3X, Inset lens: 4.0X
  • Small Base (15/8" dia.)