Pinnacle® DB C18 HPLC Columns (USP L1), Restek
Supplier: Restek
9414212 9414252 9414312 9414315 9414332 9414352 9414355 9414512 9414515 9414552 9414555 9414562 9414565 9414572 9414575 941431E 941435E 941451E 941456E 941457E 9414522 9414525 9414532 9414535 941433E 9414355-700 941452E 941453E 9414555-700 941455E 9414565-700 9414575-700 9414232 9414335
10059-864EA 994.4 USD
10059-864 10059-866 10059-868 10059-870 10059-872 10059-876 10059-878 10059-880 10059-882 10059-884 10059-886 10059-888 10059-890 10059-892 10059-894 10060-122 10060-124 10060-126 10060-128 10060-130 10837-668 10837-670 10837-674 10837-676 10838-532 10838-534 10838-536 10838-538 10838-540 10838-542 10838-544 10838-546 10838-548 10838-550 10838-552 10838-554 10838-556 10838-558 10838-560 10838-562 10838-564 10838-566 10838-568 10838-570 10838-572 10838-574 10838-576 10838-578 10838-580 10838-582 10838-584 10838-586 10838-588 10838-590 10838-592 10839-814 10839-820
Pinnacle® DB C18 HPLC Columns (USP L1), Restek
Chromatography Columns
The general-purpose Restek C18 is a conventional monomeric octadecylsilane column suitable for analyses of a wide range of compounds from acidic through slightly basic.
  • Suitable for a wide range of compounds
  • Optimized for UHPLC stability
  • Combines selectivity and efficiency
Pinnacle DB columns are 100% manufactured by Restek in our Bellefonte, Pennsylvania facility. Because performance begins with the support, our Pinnacle DB UHPLC columns start with base-deactivated spherical silica that is optimized for UHPLC stability. From there, we bond them with a wide variety of phases to give chromatographers a stable and selective column. Get the most out of your UHPLC system. Combine selectivity and efficiency by using Restek Pinnacle DB UHPLC columns.