Resprep® Oil and Grease SPE Disks, Restek
Supplier: Restek
26022
10059-482PK 244.39 USD
10059-482
Resprep® Oil and Grease SPE Disks, Restek
Filters Filter Discs
Disks reduce emulsion formation and the amount of solvent required by previous EPA methods.
  • Reduce the amount of solvent required by previous EPA methods
  • Reduced emulsion formation
  • Disks are embedded with specialty bonded silica
These 47 mm glass fiber disks are embedded with specialty bonded silica. No chlorofluorocarbons are needed. Disks meet requirements for EPA method 1664 (a sodium sulfate drying tube and a 0.45 μm PTFE syringe filter may also be used).
Certifications: Disks meet requirements for EPA method 1664.