Gas-Tight Syringe Replacement Needles for Luer Lock Syringes, SGE, Restek
Supplier: Restek
24763
10059-262PK 96.09 USD
10059-262 10059-264
Gas-Tight Syringe Replacement Needles for Luer Lock Syringes, SGE, Restek
Syringe Needles
For use with Luer lock syringes.
  • Compatible with syringe volumes of 1 – 100 mL