Micro-Volume, Positive Displacement Syringes, Hamilton, Restek
Supplier: Restek
24551
10059-194EA 198.92 USD
10059-194
Micro-Volume, Positive Displacement Syringes, Hamilton, Restek
Syringes Syringes for Chromatography
Sample is contained in the needle with full plunger displacement, ensuring no dead volume.
  • High precision and accuracy down to 0.1 µL
  • Replaceable needle/plunger kits and barrels (available on request)
Seal between the plunger and the needle can be tightened for high-pressure injection.